Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

336068

Sigma-Aldrich

2,5-Dimethyl-1,4-phenylenediamine

97%

Synonym(s):

2,5-Dimethyl-1,4-diaminobenzene, 2,5-Dimethyl-p -phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H2(NH2)2
CAS Number:
Molecular Weight:
136.19
EC Number:
MDL number:
UNSPSC Code:
12162002
eCl@ss:
39030503
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

powder

mp

149-150 °C (lit.)

SMILES string

Cc1cc(N)c(C)cc1N

InChI

1S/C8H12N2/c1-5-3-8(10)6(2)4-7(5)9/h3-4H,9-10H2,1-2H3

InChI key

BWAPJIHJXDYDPW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Zhang et al.
Analytical methods : advancing methods and applications, 13(1), 8-33 (2020-11-28)
Porous organic frameworks (POFs) are a kind of porous material with a network structure composed of repeated monomers, which have excellent physical and chemical properties, such as a high surface area, high porosity, uniform pore sizes and structural diversity, and
Xing Han et al.
Chemical Society reviews, 49(17), 6248-6272 (2020-07-30)
Covalent organic frameworks (COFs) are constructed using reticular chemistry with the building blocks being connected via covalent bonds and have emerged as a new series of porous materials for multitudinous applications. Most COFs reported to date are achiral, and only
Erin L Willis et al.
PloS one, 6(7), e21159-e21159 (2011-07-19)
After ovulation, non-pregnant female giant pandas experience pseudopregnancy. During pseudopregnancy, non-pregnant females exhibit physiological and behavioral changes similar to pregnancy. Monitoring hormonal patterns that are usually different in pregnant mammals are not effective at determining pregnancy status in many animals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service