Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

335312

Sigma-Aldrich

2-Pentyn-1-ol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C≡CCH2OH
CAS Number:
Molecular Weight:
84.12
Beilstein:
1734007
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.452 (lit.)

bp

84-85 °C/57 mmHg (lit.)

density

0.909 g/mL at 25 °C (lit.)

SMILES string

CCC#CCO

InChI

1S/C5H8O/c1-2-3-4-5-6/h6H,2,5H2,1H3

InChI key

WLPYSOCRPHTIDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Pentyn-1-ol was employed as starting reagent for the synthesis of (-)-muricatacin. It was also used in the preparation of (2Z)-3-tributylstannyl-2-penten-1-ol.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Wada et al.
Chemical & pharmaceutical bulletin, 48(9), 1391-1394 (2000-09-19)
Palladium catalyzed cross coupling reactions of a vinyl triflate intermediate and various alkenyl stannanes afforded trisubstituted Z-olefins stereoselectively in high yields. These olefins were then converted to the corresponding 9Z-retinoic acids via Horner-Emmons reaction and subsequent basic hydrolysis in excellent
H Makabe et al.
Bioscience, biotechnology, and biochemistry, 57(6), 1028-1029 (1993-06-01)
The synthesis of (-)-muricatacin starting from 1-bromododecane and 2-pentyn-1-ol is described. 2-Pentadecyn-1-ol (4), which was prepared from 1-bromododecane (2) and 2-pentyn-1-ol (3), was converted to epoxy alcohol 6 through a two-step reaction sequence, 6 being successively submitted to tosylation, iodination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service