Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

332046

Sigma-Aldrich

Ethylenediaminetetraacetic dianhydride

98%

Synonym(s):

4,4′-Ethylenebis(2,6-morpholinedione), EDTA dianhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O6
CAS Number:
Molecular Weight:
256.21
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

mp

190 °C (dec.) (lit.)

SMILES string

O=C1CN(CCN2CC(=O)OC(=O)C2)CC(=O)O1

InChI

1S/C10H12N2O6/c13-7-3-11(4-8(14)17-7)1-2-12-5-9(15)18-10(16)6-12/h1-6H2

InChI key

POLIXZIAIMAECK-UHFFFAOYSA-N

General description

Ethylenediaminetetraacetic dianhydride (EDTAD) is a ramification of ethylenediaminetetraacetate (EDTA), which consists of two anhydride groups that can react with the hydroxyl and the amino groups. It is a biodegradable compound that is used as a chelating agent.

Application

  • Electrospun cellulose acetate/activated carbon composite modified by EDTA: This study explores the application of ethylenediaminetetraacetic acid dianhydride in creating efficient dye removal systems from water. The modification led to significant improvements in adsorption properties (Elmaghraby et al., 2023).
  • Application of a new chitosan derivative for adsorption of copper (II) and oxyanions of chromium (VI): This research describes the use of ethylenediaminetetraacetic dianhydride as a modifier in creating bioadsorbents from chitosan, targeting heavy metal and dye removal from wastewater (Lucena de Sousa et al., 2024).
  • Lignocellulosic biomass functionalized with EDTA dianhydride for removing Cu (II) and dye from wastewater: This article discusses the modification of Gundelia Tournefortii with ethylenediaminetetraacetic dianhydride to enhance its capabilities for copper and dye adsorption in wastewater treatment processes (Safardastgerdi et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study on the adsorption of Ca2+, Cd2+ and Pb2+ by magnetic Fe3O4 yeast treated with EDTA dianhydride
Xu M, et al.
Chemical Engineering Journal, 168(2), 737-745 (2011)
Novel amphoteric pH-sensitive hydrogels derived from ethylenediaminetetraacetic dianhydride, butanediamine and amino-terminated poly (ethylene glycol): Design, synthesis and swelling behavior
Luo Y, et al.
European Polymer Journal, 47(1), 40-47 (2011)
Synthesis and characterization of a novel versatile poly (amic acid) derived from ethylenediaminetetraacetic dianhydride
Padavan DT and Wan WK
Materials Chemistry and Physics, 124(1), 427-433 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service