Skip to Content
MilliporeSigma
All Photos(1)

Documents

329991

Sigma-Aldrich

2-Chloromethyl-1,3-dioxolane

97%

Synonym(s):

Chloroacetaldehyde ethylene acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7ClO2
CAS Number:
Molecular Weight:
122.55
Beilstein:
103229
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.449 (lit.)

bp

157-158 °C (lit.)

density

1.234 g/mL at 25 °C (lit.)

SMILES string

ClCC1OCCO1

InChI

1S/C4H7ClO2/c5-3-4-6-1-2-7-4/h4H,1-3H2

InChI key

IKZOMJGRWIOEDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloromethyl-1,3-dioxolane is commonly used in industrial manufacturing processes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Luong et al.
Journal of chromatography. A, 1255, 216-220 (2012-03-14)
Oxygenated compounds like methanol, ethanol, 1-propanol, 2-propanol, 1-butanol, acetaldehyde, crotonaldehyde, ethylene oxide, tetrahydrofuran, 1,4-dioxane, 1,3-dioxolane, and 2-chloromethyl-1,3-dioxolane are commonly encountered in industrial manufacturing processes. Despite the availability of a variety of column stationary phases for chromatographic separation, it is difficult

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service