Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

328901

Sigma-Aldrich

2-Nitro-4-(trifluoromethyl)benzoic acid

98%

Synonym(s):

2-Nitro-α,α,α-trifluor-p-toluic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(CF3)CO2H
CAS Number:
Molecular Weight:
235.12
Beilstein:
1985588
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

134-138 °C (lit.)

functional group

carboxylic acid
fluoro
nitro

SMILES string

OC(=O)c1ccc(cc1[N+]([O-])=O)C(F)(F)F

InChI

1S/C8H4F3NO4/c9-8(10,11)4-1-2-5(7(13)14)6(3-4)12(15)16/h1-3H,(H,13,14)

InChI key

MYSAXQPTXWKDPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Nitro-4-(trifluoromethyl)benzoic acid was used in preparation of 4-trifluoromethyl anthranilic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Przemysław Szczeciński et al.
Acta biochimica Polonica, 55(4), 749-752 (2008-11-29)
Organic extracts of six urine samples from children treated with nitisinone, a medicine against tyrosinemia type I, were investigated by (1)H and (19)F NMR spectroscopy. The presence of unchanged 2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione (NTBC), 6-hydroxy-2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione (NTBC-OH) and 2-nitro-4-trifluoromethylbenzoic acid (NTFA) as well as
Hanna Barchanska et al.
Journal of pharmaceutical and biomedical analysis, 171, 15-21 (2019-04-09)
Nitisinone (2-(2-nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, NTBC) was the first synthetically produced triketone herbicide. However, its unsatisfactory herbicidal properties, negative impact on the natural environment and the high cost of synthesis have hindered its commercialization as a plant protection agent. Nevertheless, NTBC has become

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service