Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324663

Sigma-Aldrich

1,4-Pentadien-3-ol

contains 0.4% hydroquinone as stabilizer, ≥96%

Synonym(s):

Divinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH(OH)CH=CH2
CAS Number:
Molecular Weight:
84.12
Beilstein:
1735809
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96%

form

liquid

contains

0.4% hydroquinone as stabilizer

refractive index

n20/D 1.445 (lit.)

bp

115-116 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OC(C=C)C=C

InChI

1S/C5H8O/c1-3-5(6)4-2/h3-6H,1-2H2

InChI key

ICMWSAALRSINTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for asymmetric epoxidation.
Substrate employed in a synthesis of amino-substituted dienes via a bismuth-catalyzed SNi displacement of alcohols by sulfonamide nucleophiles.
Useful building block in natural product synthesis.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Asymmetry, 4, 1533-1533 (1993)
Masanori Imai et al.
The Journal of organic chemistry, 69(4), 1144-1150 (2004-02-14)
Intermolecular hydroacylation between salicylaldehydes 1, 26-40 and 1,4-penta- or 1,5-hexadienes 4-13 by Rh-catalyst proceeded under mild reaction conditions to give a mixture of iso- and normal-hydroacylated products 14-25, 41-55, and 57-60. In the hydroacylation reaction, chelation of both salicylaldehyde and
P Andrew Evans et al.
Journal of the American Chemical Society, 125(48), 14702-14703 (2003-12-04)
The enantioselective total synthesis of the annonaceous acetogenin (-)-mucocin (1) was accomplished using a triply convergent 12-step sequence (longest linear sequence) in 13.6% overall yield. This represents the first application of the temporary silicon-tethered (TST) ring-closing metathesis (RCM) cross-coupling reaction
Chin. J. Chem., 9, 381-381 (1991)
Bismuth-catalyzed direct substitution of the hydroxy group in alcohols with sulfonamides, carbamates, and carboxamides.
Hongbo Qin et al.
Angewandte Chemie (International ed. in English), 46(3), 409-413 (2006-12-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service