Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

319708

Sigma-Aldrich

Cyclopentanethiol

97%

Synonym(s):

Cyclopentyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9SH
CAS Number:
Molecular Weight:
102.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

0.96 (vs air)

vapor pressure

1 mmHg ( 20 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.4902 (lit.)

bp

129-131 °C/745 mmHg (lit.)

density

0.955 g/mL at 25 °C (lit.)

functional group

thiol

SMILES string

SC1CCCC1

InChI

1S/C5H10S/c6-5-3-1-2-4-5/h5-6H,1-4H2

InChI key

WVDYBOADDMMFIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service