Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

311006

Sigma-Aldrich

Potassium chromium(III) oxalate trihydrate

98%

Synonym(s):

Potassium tris(oxalato)chromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K3Cr(C2O4)3·3H2O
CAS Number:
Molecular Weight:
487.39
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystalline

grade

for analytical purposes

Assay

98%

form

crystalline

SMILES string

O.O.O.[K+].[K+].[K+].[O-]C(=O)C(=O)O[Cr](OC(=O)C([O-])=O)OC(=O)C([O-])=O

InChI

1S/3C2H2O4.Cr.3K.3H2O/c3*3-1(4)2(5)6;;;;;;;/h3*(H,3,4)(H,5,6);;;;;3*1H2/q;;;+3;3*+1;;;/p-6

InChI key

OPOKJTVMNOKKQO-UHFFFAOYSA-H

Related Categories

Application

Used to prepare Langmuir-Blodgett thin films which have magnetic properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E Alegria et al.
Biochemistry, 28(21), 8653-8658 (1989-10-17)
The radicals generated by adriamycin-sensitive (CHO-AB) and adriamycin-resistant (CHO-C5) Chinese hamster ovary cells as well as by adriamycin-sensitive and -resistant human breast cancer cells (MCF7-WT and MCF7-ADR) have been studied with spin-trapping and ESR spectroscopy. During anoxic exposure to adriamycin
Z Heng et al.
Wei sheng yan jiu = Journal of hygiene research, 30(3), 149-151 (2003-01-15)
Apoptosis of immature thymocyte of mice induced by dexamethasone and DNA strand-broken cells induced by K2Cr2O7 were tested by comet assay. The results showed that the morphology of apoptotic cells assayed by comet was special. Under the same condition, the
K Anzai et al.
Biochimica et biophysica acta, 937(1), 73-80 (1988-01-13)
We have developed an electron spin resonance (ESR)-stopped flow technique and employed it for the simple and rapid determination of internal volumes of biomembrane vesicles and liposomes. A vesicle suspension containing a neutral and membrane-permeable spin label, 2,2,6,6-tetramethyl-4-oxopiperidine-1-oxyl (TEMPONE), was
G L Rikken et al.
Nature, 405(6789), 932-935 (2000-07-06)
Many chemical and physical systems can occur in two forms distinguished solely by being mirror images of each other. This phenomenon, known as chirality, is important in biochemistry, where reactions involving chiral molecules often require the participation of one specific
Chirality, magnetism and light
L D Barron
Nature, 405(6789), 895-896 (2000-07-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service