Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

309192

Sigma-Aldrich

Rhenium(V) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ReCl5
CAS Number:
Molecular Weight:
363.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystalline

form

crystalline

Quality Level

reaction suitability

reagent type: catalyst
core: rhenium

concentration

50.0-53.0% Re (With Tetraphenylarsonium Chloride, gravimetric)

SMILES string

Cl[Re](Cl)(Cl)(Cl)Cl

InChI

1S/5ClH.Re/h5*1H;/q;;;;;+5/p-5

InChI key

XZQYTGKSBZGQMO-UHFFFAOYSA-I

Application

Starting material for rhenium porphyrins and alkyne complexes of rhenium.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Vrabec et al.
Ceskoslovenska pediatrie, 45(9), 518-519 (1990-09-01)
The authors evaluate the effect of hormonal treatment with chorionic gonadotropin on histological structures of the testes in experimental work and in artificially induced cryptorchism in the rabbit; they evaluate histological findings in cryptorchid and normally descended testes.
F Decian et al.
Minerva chirurgica, 45(19), 1227-1237 (1990-10-15)
Hyperthermic antiblastic isolated perfusion is a method largely used for the treatment of locally advanced limb melanoma. The method requires vascular isolation and hyperthermic perfusion of the limb using an extracorporeal circuit and administering the melphalan as antiblastic drug. Twenty-six

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service