308595
2-Bromopropionamide
99%
Synonym(s):
α-Bromopropanamide, α-Bromopropionamide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3CH(Br)CONH2
CAS Number:
Molecular Weight:
151.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
mp
123-125 °C (lit.)
functional group
amide
bromo
SMILES string
CC(Br)C(N)=O
InChI
1S/C3H6BrNO/c1-2(4)3(5)6/h2H,1H3,(H2,5,6)
InChI key
AUHYZQCEIVEMFH-UHFFFAOYSA-N
General description
The hydrolytic dehalogenation of 2-bromopropionamide was catalyzed by dl-2-haloacid dehalogenase from Pseudomonas putida PP3 (dl-DEX 312).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A new dl-2-haloacid dehalogenase acting on 2-haloacid amides: purification, characterization, and mechanism.
Journal of Molecular Catalysis. B, Enzymatic, 23(2), 329-336 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service