Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

304182

Sigma-Aldrich

cis-2-Penten-1-ol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH2OH
CAS Number:
Molecular Weight:
86.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

95%

form

liquid

refractive index

n20/D 1.437 (lit.)

bp

138 °C (lit.)

density

0.853 g/mL at 25 °C (lit.)

SMILES string

[H]\C(CC)=C(/[H])CO

InChI

1S/C5H10O/c1-2-3-4-5-6/h3-4,6H,2,5H2,1H3/b4-3-

InChI key

BTSIZIIPFNVMHF-ARJAWSKDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

cis-2-Penten-1-ol is also known as banana flavoured compound. It was found to reduce the efficiency of sex pheromone lures in trapping male moths in a proof long-range field trapping experiment. Kinetics of gas-phase reactions of NO3 radical with cis-2-penten-1-ol was investigated using relative rates technique.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhijun Zhao et al.
The journal of physical chemistry. A, 115(44), 12161-12172 (2011-10-15)
The gas-phase reactions of NO(3) radicals with series of 1-alkenes, dienes, cycloalkenes, alkenols, and alkenals were studied in pure N(2) or 20% O(2)/80% N(2) bath gas at room temperature and atmospheric pressure using a relative rates technique. Rate coefficients were
Rui Tang et al.
PloS one, 7(11), e49256-e49256 (2012-11-21)
Volatile organic compounds (VOCs) were collected from damaged and intact mulberry leaves (Morus alba L., Moraceae) and from Hyphantria cunea larvae by headspace absorption with Super Q columns. We identified their constituents using gas chromatography-mass spectrometry, and evaluated the responses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service