Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

304077

Sigma-Aldrich

3-Methylcrotonaldehyde

97%

Synonym(s):

3,3-Dimethylacrolein, 3,3-Dimethylacrylaldehyde, NSC 149164, Prenal, Senecialdehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCHO
CAS Number:
Molecular Weight:
84.12
Beilstein:
1734740
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

7 mmHg ( 20 °C)

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.462 (lit.)
n20/D 1.462

bp

133-135 °C (lit.)

solubility

water: soluble 110 g/L at 20 °C

density

0.878 g/mL at 20 °C
0.872 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)\C=C(\C)C

InChI

1S/C5H8O/c1-5(2)3-4-6/h3-4H,1-2H3

InChI key

SEPQTYODOKLVSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methylcrotonaldehyde is an α, β-unsaturated aldehyde. Electrochemical dimerization of 3-methylcrotonaldehyde was reported. The hydrogenation of 3-methylcrotonaldehyde over Ru, Pt and Rh was investigated.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service