Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

303054

Sigma-Aldrich

2′-Hydroxy-1′-acetonaphthone

99%

Synonym(s):

1-Acetyl-2-naphthol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC10H6COCH3
CAS Number:
Molecular Weight:
186.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

65-67 °C (lit.)

functional group

ketone

SMILES string

CC(=O)c1c(O)ccc2ccccc12

InChI

1S/C12H10O2/c1-8(13)12-10-5-3-2-4-9(10)6-7-11(12)14/h2-7,14H,1H3

InChI key

VUIOUIWZVKVFCI-UHFFFAOYSA-N

Application

2′-Hydroxy-1′-acetonaphthone was used in preparation of 1-(N-isopropyl-a-iminoethyl)-2-naphthol, Schiff base.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intramolecular hydrogen bond investigations in Schiff bases derivatives of 2-hydroxy-1-naphthaldehyde and 2-hydroxy-1-acetonaphthone in CDCl< sub> 3</sub> solution and in the solid state by NMR methods.
Schilf W, et al.
Journal of Molecular Structure, 602, 41-47 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service