Skip to Content
MilliporeSigma

299367

Urea-12C

99.9 atom % 12C

Synonym(s):

Carbamide-12C

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
H2N12CONH2
CAS Number:
Molecular Weight:
60.04
UNSPSC Code:
12352111
PubChem Substance ID:
EC Number:
200-315-5
MDL number:
eCl@ss:
32150917
Isotopic purity:
99.9 atom % 12C
Mass shift:
depleted
Form:
solid


description

13C-depleted

Quality Segment

isotopic purity

99.9 atom % 12C

form

solid

mp

132-135 °C (lit.)

mass shift

depleted

SMILES string

N[12C](N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)/i1+0

InChI key

XSQUKJJJFZCRTK-IGMARMGPSA-N

Gene Information

human ... CA1(759), CA2(760), EPHX2(2053)
mouse ... Ephx2(13850)
rat ... Ppm1a(24666)

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact
Stable Isotopes Customer Service.



Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library