Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

298689

Sigma-Aldrich

DL-Isoleucine

ReagentPlus®, 99%

Synonym(s):

DL-2-Amino-3-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH(NH2)COOH
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721790
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

reaction suitability

reaction type: solution phase peptide synthesis

mp

290 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CC[C@@H](C)[C@@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m1/s1

InChI key

AGPKZVBTJJNPAG-RFZPGFLSSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A mixture of stereoisomers.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service