Skip to Content
MilliporeSigma
All Photos(1)

Documents

296678

Sigma-Aldrich

(S)-(−)-1,2,4-Butanetriol

98%

Synonym(s):

(2S)-(-)-Butane-1,2,4-triol, (S)-(-)-1,2,4-Trihydroxybutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
106.12
Beilstein:
1719408
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]19/D −28±2, c = 1 in methanol

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.475 (lit.)

bp

150 °C/0.04 mmHg (lit.)

density

1.19 g/mL at 25 °C (lit.)

SMILES string

OCC[C@H](O)CO

InChI

1S/C4H10O3/c5-2-1-4(7)3-6/h4-7H,1-3H2/t4-/m0/s1

InChI key

ARXKVVRQIIOZGF-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(-)-1,2,4-Butanetriol can be prepared via reduction of (S)-malic acid in the presence of borane-dimethyl sulfide.

Application

(S)-(-)-1,2,4-Butanetriol may be used as a starting material in the enantioselective total syntheses of (+)-azimine and (+)-carpaine.
It can also be used to prepare the following organic building blocks:
  • (+)-3,4-epoxy-1-butanol
  • (2S,4S)-4-(hydroxymethyl)-2-ferrocenyl-1,3-dioxane
  • (S)-1,2,4-triacetoxybutane via acetylation with acetic anhydride
  • (S)-1,2,4-tris-(3,5-dinitrobenzoy1oxy)butane via esterification with 3,5-dinitrobenzoyl chloride

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An efficient asymmetric synthesis of 2-substituted ferrocenecarboxaldehydes
Riant O, et al
The Journal of Organic Chemistry, 62(20), 6733-6745 (1997)
Asymmetric Michael addition of thiophenol to maleic acid esters
Yamashita H and Mukaiyama T
Chemistry Letters (Jpn), 14(3), 363-366 (1985)
Facile access to (S)-1, 2, 4-butanetriol and its derivatives.
Hanessian S, et al.
Canadian Journal of Chemistry, 62(11), 2146-2147 (1984)
Enantioselective total synthesis of (+)-azimine and (+)-carpaine.
Sato T, et al.
Organic Letters, 5(21), 3839-3842 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service