Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

293180

Sigma-Aldrich

Triethyl phosphonoacetate-1-13C

99 atom % 13C

Synonym(s):

Ethyl 2-(diethoxyphosphinyl)acetate-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(O)CH213CO2C2H5
CAS Number:
Molecular Weight:
225.18
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

reaction suitability

reaction type: C-C Bond Formation
reaction type: Olefinations

refractive index

n20/D 1.431 (lit.)

bp

142-145 °C/9 mmHg (lit.)

density

1.13 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

CCO[13C](=O)CP(=O)(OCC)OCC

InChI

1S/C8H17O5P/c1-4-11-8(9)7-14(10,12-5-2)13-6-3/h4-7H2,1-3H3/i8+1

InChI key

GGUBFICZYGKNTD-VJJZLTLGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service