Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

288462

Sigma-Aldrich

Phosphorus tribromide

≥99.99% trace metals basis

Synonym(s):

Phosphorus(III) bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PBr3
CAS Number:
Molecular Weight:
270.69
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.27 psi ( 54 °C)

Assay

≥99.99% trace metals basis

form

liquid

refractive index

n20/D 1.697 (lit.)

bp

175 °C (lit.)

mp

−41.5 °C (lit.)

density

2.88 g/mL at 20 °C (lit.)

SMILES string

BrP(Br)Br

InChI

1S/Br3P/c1-4(2)3

InChI key

IPNPIHIZVLFAFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphorus tribromide (PBr3) is a phosphorus halide that can be used as a reactive phosphorylating agent for the formation of multifunctional thiophenes.

Application

PBr3 can be used as an acceptor molecule in the formation of donor-acceptor complexes. It can also be used in the phosphorylation of arylphospholes which can further be utilized in the synthesis of phosphonic amides.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorylation of 2-(3-methyl-1, 3-diazabuten-1-yl)-3-ethoxycarbonylthiophenes with phosphorus (III) halides
Pinchuk AM, et al.
Heteroatom Chem., 12(7), 641-651 (2001)
Site-selective phosphorylation of arylphospholes through reaction with phosphorus tribromide
Keglevich G, et al.
Journal of Organometallic Chemistry, 643(11), 32-38 (2002)
Donor-acceptor complexes between organoamines and phosphorus tribromide
Muller G, et al.
Zeitschrift fur Naturforschung B, 56(11), 1163-1171 (2001)
Andrey S Mereshchenko et al.
Nature chemistry, 7(7), 562-568 (2015-06-24)
'Roaming' is a new and unusual class of reaction mechanism that has recently been discovered in unimolecular dissociation reactions of isolated molecules in the gas phase. It is characterized by frustrated bond cleavage, after which the two incipient fragments 'roam'

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service