Skip to Content
MilliporeSigma
All Photos(2)

Documents

281522

Sigma-Aldrich

Montmorillonite K 10

Synonym(s):

MMT, MMT-K10, Montmorillonite, Montmorillonite KSF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

form

powder

Quality Level

surface area

220-270 m2/g

InChI

1S/2Al.4O2Si.H2O.3O/c;;4*1-3-2;;;;/h;;;;;;1H2;;;/q2*+3;;;;;;3*-2

InChI key

GUJOJGAPFQRJSV-UHFFFAOYSA-N

Application

Used for:
  • Support for catalysts
  • Preparation of clay nanopaper composites of nacre-like structure

Catalyst for:
  • Conversion of triose sugars with alcohols to alkyl lactates
  • Deprotection of ketone dimethylhydrazones
Clay catalyst used in a microwave-assisted synthesis of pyrazoles from enaminones.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis, 2485-2485 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service