Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

280933

Sigma-Aldrich

(Triphenylphosphoranylidene)acetaldehyde

95%

Synonym(s):

(Formylmethylene)triphenylphosphorane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOCH=P(C6H5)3
CAS Number:
Molecular Weight:
304.32
Beilstein:
523797
EC Number:
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

185-188 °C (lit.)

functional group

aldehyde
phosphine

SMILES string

[H]C(=O)C=P(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C20H17OP/c21-16-17-22(18-10-4-1-5-11-18,19-12-6-2-7-13-19)20-14-8-3-9-15-20/h1-17H

InChI key

CQCAYWAIRTVXIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Wittig reagent for the conversion of aldehydes and ketones to two-carbon homologated, α,β-unsaturated carbonyl compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian Wang et al.
Organic letters, 9(18), 3703-3706 (2007-08-19)
The first total synthesis of (-)-aplysiallene has been completed in 16 steps and features a key sequential Mukaiyama aerobic oxidative cyclization to prepare the fused bis-THF core. The original stereochemical assignment has been revised as shown.
Tohru Nagamitsu et al.
The Journal of organic chemistry, 72(8), 2744-2756 (2007-03-16)
The total synthesis of borrelidin has been achieved. The best feature of our synthetic route is macrocyclization at C11-C12 for the construction of an 18-membered ring after esterification between two segments. A detailed examination of the macrocyclization led us to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service