Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

278777

Sigma-Aldrich

Tributylsilane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SiH
CAS Number:
Molecular Weight:
200.44
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

225-226 °C/755 mmHg (lit.)

density

0.779 g/mL at 25 °C (lit.)

SMILES string

CCCC[SiH](CCCC)CCCC

InChI

1S/C12H28Si/c1-4-7-10-13(11-8-5-2)12-9-6-3/h13H,4-12H2,1-3H3

InChI key

HIAZFYQNGXRLTF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stuart A Bartlett et al.
Journal of the American Chemical Society, 141(29), 11471-11480 (2019-07-16)
Complexes with weakly coordinating ligands are often formed in chemical reactions and can play key roles in determining the reactivity, particularly in catalytic reactions. Using time-resolved X-ray absorption fine structure (XAFS) spectroscopy in combination with time-resolved IR (TRIR) spectroscopy and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service