Skip to Content
MilliporeSigma
All Photos(1)

Documents

278351

Sigma-Aldrich

Isobutyl vinyl ether

contains 0.1% potassium hydroxide as stabilizer, 99%

Synonym(s):

Vinyl isobutyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2OCH=CH2
CAS Number:
Molecular Weight:
100.16
Beilstein:
1560159
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

3.45 (vs air)

Quality Level

vapor pressure

68 mmHg ( 20 °C)

Assay

99%

form

liquid

contains

0.1% potassium hydroxide as stabilizer

refractive index

n20/D 1.395 (lit.)

bp

82-83 °C (lit.)

mp

−112 °C (lit.)

density

0.768 g/mL at 25 °C (lit.)

SMILES string

CC(C)COC=C

InChI

1S/C6H12O/c1-4-7-5-6(2)3/h4,6H,1,5H2,2-3H3

InChI key

OZCMOJQQLBXBKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isobutyl vinyl ether (IBVE) is a vinyl ether monomer that finds wide application in the polymer industry. It is utilized in various applications such as adhesives, coatings, sealants, ink production, elastomers, agricultural chemicals, and specialty polymer formulations due to its desirable properties, including high reactivity, oxygen permeability, good solvency, and crosslinking capability. It is also used in various Biomedical Applications which include, drug delivery systems, tissue engineering, and biocompatible coatings.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher L Rock et al.
Dalton transactions (Cambridge, England : 2003), 48(2), 461-467 (2018-11-30)
The phosphine-substituted α-diimine Ni precursor, (Ph2PPrDI)Ni, has been found to catalyze alkene hydrosilylation in the presence of Ph2SiH2 with turnover frequencies of up to 124 h-1 at 25 °C (990 h-1 at 60 °C). Moreover, the selective hydrosilylation of allylic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service