Skip to Content
MilliporeSigma
All Photos(1)

Documents

277614

Sigma-Aldrich

Diazald®-(N-methyl-13C)

99 atom % 13C, 97% (CP)

Synonym(s):

N-Methyl-13C-N-nitroso-p-toluenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2N(13CH3)NO
CAS Number:
Molecular Weight:
215.23
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

solid

mp

61-62 °C (lit.)

mass shift

M+1

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)N([13CH3])N=O

InChI

1S/C8H10N2O3S/c1-7-3-5-8(6-4-7)14(12,13)10(2)9-11/h3-6H,1-2H3/i2+1

InChI key

FFKZOUIEAHOBHW-VQEHIDDOSA-N

Features and Benefits

Convenient source of diazomethane-13C.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Legal Information

Diazald is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Self-react. C - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service