Skip to Content
MilliporeSigma
All Photos(1)

Documents

277282

Sigma-Aldrich

1,1-Dimethylpropylmagnesium chloride solution

1.0 M in diethyl ether

Synonym(s):

(1,1-Dimethylpropyl)magnesium chloride, Chloro(1,1-dimethylpropyl)magnesium, tert-Amylmagnesium chloride, tert-Pentyl magnesium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)2MgCl
CAS Number:
Molecular Weight:
130.90
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in diethyl ether

density

0.831 g/mL at 25 °C

SMILES string

CCC(C)(C)[Mg]Cl

InChI

1S/C5H11.ClH.Mg/c1-4-5(2)3;;/h4H2,1-3H3;1H;/q;;+1/p-1

InChI key

NWFDYJDCSIHFAB-UHFFFAOYSA-M

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service