Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

276170

Sigma-Aldrich

N,N-Dimethylbenzamide

99%

Synonym(s):

DMBZ

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CON(CH3)2
CAS Number:
Molecular Weight:
149.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

132-133 °C/15 mmHg (lit.)

mp

43-45 °C (lit.)

SMILES string

CN(C)C(=O)c1ccccc1

InChI

1S/C9H11NO/c1-10(2)9(11)8-6-4-3-5-7-8/h3-7H,1-2H3

InChI key

IMNDHOCGZLYMRO-UHFFFAOYSA-N

General description

N,N-Dimethylbenzamide is a hydrotropic agent and its ability to solubilize drugs with low aqueous solubility has been investigated. It reacts with PhLnI complexes (Ln-Eu, Sm, Yb) to yield benzophenone in good yields. Deuterium exchange labelling experimets on N,N-dimethylbenzamide using [IrH2(Me2CO)2(PPh3)2]BF4 as catalyst and deuterium gas as the source of isotope has been conducted.

Application

Reagent for deoxygenation of secondary alcohols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deuterium exchange labelling of substituted aromatics using [IrH2 (Me2CO) 2 (PPh3)2] BF4.
Heys JR, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 33(5), 431-438 (1993)
L R Hall et al.
The Journal of biological chemistry, 265(21), 12349-12355 (1990-07-25)
Liver microsomal cytochrome P-450 readily N-dealkylates N,N-dimethylamides. N-Methyl-N-hydroxymethyl amides were isolated as intermediates and characterized by gas chromatography-mass spectrometry as their trimethylsilyl ethers. Intramolecular kinetic deuterium isotope effects measured for the enzymic N-demethylation of a series of 12 aromatic and
Ji Young Kim et al.
Journal of controlled release : official journal of the Controlled Release Society, 152(1), 13-20 (2011-03-01)
Polymer micelles have been used widely for delivery of poorly water-soluble drugs. Such drug delivery, however, has been based primarily on hydrophobic interactions. For better drug loading and improved stability, hydrotropic polymer micelles were used. To develop a versatile polymer
Reactions of ethyl-and phenyllanthanide s complexes with n, n-dimethylbenzamide and benzaldehyde.
Yokoo K, et al.
Polyhedron, 2(10), 1101-1102 (1983)
Ji Young Kim et al.
Journal of pharmaceutical sciences, 99(9), 3953-3965 (2010-07-08)
The solubilizing ability of two aromatic hydrotropes, N,N-diethylnicotinamide (DENA) and N,N-dimethylbenzamide (DMBA), was investigated using a set of 13 poorly soluble, structurally diverse drugs. The number of aromatic rings in the solute molecule has a very strong effect on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service