Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

275867

Sigma-Aldrich

5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine cobalt(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H36CoN4O4
CAS Number:
Molecular Weight:
791.76
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

reaction suitability

reagent type: catalyst
core: cobalt

λmax

417 nm
530 nm (2nd)

SMILES string

COc1ccc(cc1)-c2c3ccc(n3)c(-c4ccc(OC)cc4)c5ccc6c(-c7ccc(OC)cc7)c8ccc(n8)c(-c9ccc(OC)cc9)c%10ccc2n%10[Co]n56

InChI

1S/C48H36N4O4.Co/c1-53-33-13-5-29(6-14-33)45-37-21-23-39(49-37)46(30-7-15-34(54-2)16-8-30)41-25-27-43(51-41)48(32-11-19-36(56-4)20-12-32)44-28-26-42(52-44)47(40-24-22-38(45)50-40)31-9-17-35(55-3)18-10-31;/h5-28H,1-4H3;/q-2;+2/b45-37-,45-38-,46-39-,46-41-,47-40-,47-42-,48-43-,48-44-;

InChI key

QBCIMRXPMLWVML-NHZJRHMYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Starting material in the synthesis of cobalt nitrosyl porphyrins for use in the study of the binding and activation of nitric oxide.

Features and Benefits

Acts as a versatile catalyst during the oxidation of a wide range of organic substrates with dioxygen in the presence of 2-methylpropanal under ambient conditions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service