Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

274755

Sigma-Aldrich

4-Nitrophthalic acid

92%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(CO2H)2
CAS Number:
Molecular Weight:
211.13
Beilstein:
1882262
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

92%

impurities

5-8% 3-nitrophthalic acid

mp

159-161 °C (lit.)

functional group

carboxylic acid
nitro

SMILES string

OC(=O)c1ccc(cc1C(O)=O)[N+]([O-])=O

InChI

1S/C8H5NO6/c10-7(11)5-2-1-4(9(14)15)3-6(5)8(12)13/h1-3H,(H,10,11)(H,12,13)

InChI key

SLBQXWXKPNIVSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The tissue-specific activity of 4-nitrophthalic acid was studied.

Application

4-Nitrophthalic acid was used in the preparation of a 2D homochiral inorganic-organic framework {[Mn(4-nitrophthalate)(4,4′-bipyridine)(H2O)]·(H2O2}n by reacting with manganese ions, and ancillary 4,4′-bipyridine ligands.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wei Li et al.
Dalton transactions (Cambridge, England : 2003), 40(27), 7147-7152 (2011-03-05)
A 2D homochiral inorganic-organic framework {[Mn(NPTA)(4,4'-bpy)(H(2)O)]·(H(2)O)(2)}(n) was prepared by assembling achiral polar 4-nitrophthalic acid, manganese ions, and ancillary 4,4'-bipyridine ligands (NPTA = 4-nitrophthalate) (4,4'-bpy = 4,4'-bipyridine). The isomorphous Zn(ii) compound was also prepared as a diamagnetic analogue. Adjacent manganese spin
Xiaoyong Wu et al.
Water environment research : a research publication of the Water Environment Federation, 85(5), 404-410 (2013-06-26)
In order to find the relationship between tissue-specific expressions of glutathione S-transferases (GSTs) and their function in preventing effects of environmental toxicants, GSTs were primarily purified by GST-Sepharose 4B affinity chromatography from liver intestine, gill, mantle, and adductor muscle of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service