Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

269239

Sigma-Aldrich

Tridecanal

technical grade, 90%

Synonym(s):

Tridecyl aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11CHO
CAS Number:
Molecular Weight:
198.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.438 (lit.)

bp

132-136 °C/8 mmHg (lit.)

density

0.835 g/mL at 25 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCC=O

InChI

1S/C13H26O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14/h13H,2-12H2,1H3

InChI key

BGEHHAVMRVXCGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The uptake of NO(3) on solid tridecanal was studied.

Application

Tridecanal was used as a model compound in the development of nonradioactive assays for the enzymes 2-hydroxyacyl-CoA lyase (HACL1) and sphingosine-1-phosphate lyase (SGPL1) employing the Hantzsch reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard Iannone et al.
Physical chemistry chemical physics : PCCP, 13(21), 10214-10223 (2011-04-22)
We report the first measurements of the reactive uptake of NO(3) with condensed-phase aldehydes. Specifically, we studied NO(3) uptake on solid tridecanal and the uptake on liquid binary mixtures containing tridecanal and saturated organic molecules (diethyl sebacate, dioctyl sebacate, and
Serena Mezzar et al.
Journal of lipid research, 55(3), 573-582 (2013-12-11)
Long-chain aldehydes are commonly produced in various processes, such as peroxisomal α-oxidation of long-chain 3-methyl-branched and 2-hydroxy fatty acids and microsomal breakdown of phosphorylated sphingoid bases. The enzymes involved in the aldehyde-generating steps of these processes are 2-hydroxyacyl-CoA lyase (HACL1)
Karolina Wieszczycka et al.
Photochemistry and photobiology, 91(4), 786-796 (2015-03-13)
The goal of the research was to study the reactivity of the hydrophobic 2- and 3-pyridineketoximes under exposure to UV-VIS light. The photodegradation was conducted in both toluene and heptane for 10 h under atmosphere of argon. Ten-hour irradiation experiments demonstrated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service