Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

268925

Sigma-Aldrich

Butyl ethyl ether

99%

Synonym(s):

1-Ethoxybutane, Ethyl butyl ether, Ethyl n-butyl ether, n-Butyl ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OC2H5
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731323
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

52 mmHg ( 25 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.382 (lit.)

bp

91-92 °C (lit.)

mp

−124 °C (lit.)

density

0.75 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CCCCOCC

InChI

1S/C6H14O/c1-3-5-6-7-4-2/h3-6H2,1-2H3

InChI key

PZHIWRCQKBBTOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

tert-Butyl ethyl ether was used in the determination of acetyl groups in proteins and peptides[1].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microenzymic method for the quantitative analysis of acetyl groups in proteins and peptides.
L D Stegink
Analytical biochemistry, 20(3), 502-516 (1967-09-01)

Questions

  1. こちらはアルゴン雰囲気での封入でしょうか。

    1 answer
    1. This product is not packaged under inert gas, including argon.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service