Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

26564

Sigma-Aldrich

Chlorotripyrrolidinophosphonium hexafluorophosphate

≥98.0% (AT)

Synonym(s):

PyCloP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H24ClN3P · PF6
CAS Number:
Molecular Weight:
421.73
Beilstein:
6842341
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (AT)

form

solid

reaction suitability

reaction type: Coupling Reactions

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.Cl[P+](N1CCCC1)(N2CCCC2)N3CCCC3

InChI

1S/C12H24ClN3P.F6P/c13-17(14-7-1-2-8-14,15-9-3-4-10-15)16-11-5-6-12-16;1-7(2,3,4,5)6/h1-12H2;/q+1;-1

InChI key

BSCYRXJVGSZNKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for:
Peptide coupling reactions and synthesis of coupling reagents
Sequence selective peptide recognition
Synthesis of condensing reagents
Synthesis of heterocyclic β-sheet ligands

Other Notes

Crystalline reagent for peptide coupling of N-alkylated N-Fmoc and N-Z amino acids in high yield and without racemization.

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Coste et al.
The Journal of Organic Chemistry, 59, 2437-2437 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service