Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

265187

Sigma-Aldrich

2,5-Difluorobenzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CHO
CAS Number:
Molecular Weight:
142.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.498 (lit.)

bp

67-69 °C/17 mmHg (lit.)

density

1.308 g/mL at 25 °C (lit.)

functional group

aldehyde
fluoro

SMILES string

Fc1ccc(F)c(C=O)c1

InChI

1S/C7H4F2O/c8-6-1-2-7(9)5(3-6)4-10/h1-4H

InChI key

VVVOJODFBWBNBI-UHFFFAOYSA-N

General description

The emission and absorption spectra of 2,5-difluorobenzaldehyde were studied.

Application

2,5-Difluorobenzaldehyde was used in the synthesis of fluorinated analog by Henry condensation with nitroethane.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectroscopy and dynamics of the excited singlet and triplet states of 2, 3-, 2, 4-and 2, 5-difluorobenzaldehyde vapors.
Itoh, T.
Journal of Molecular Spectroscopy, 257(2), 170-176 (2009)
Matthew A Parker et al.
Bioorganic & medicinal chemistry, 16(8), 4661-4669 (2008-02-26)
The lipophilicity of a set of 5-HT(2A) ligands was determined using immobilized-artificial-membrane chromatography, a method that generates values well correlated with octanol-water partition coefficients. For agonists, a highly significant linear correlation was observed between binding affinity and lipophilicity. For ligands

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service