Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

262064

Sigma-Aldrich

mono-Bromoborane methyl sulfide complex solution

1.0 M in methylene chloride

Synonym(s):

Bromo(dimethyl sulfide)borane, Bromo(dimethyl sulfide)dihydroboron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S·BH2Br
CAS Number:
Molecular Weight:
154.86
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

1.0 M in methylene chloride

density

1.347 g/mL at 25 °C

functional group

thioether

SMILES string

BBr.CSC

InChI

1S/C2H6S.BBrH2/c1-3-2;1-2/h1-2H3;1H2

InChI key

AQRNIOMHNXJPFD-UHFFFAOYSA-N

Application

Reactant for:
  • Substitution reactions for synthesis of N-heterocyclic carbene boranes with boron-heteroatom bonds
  • Hydroboration reactions
  • Regio- and chemoselective brominative cleavage of terminal epoxides

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

50.0 °F

Flash Point(C)

10 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Roy, C. D.; Brown, H. C.
Australian Journal of Chemistry, 60, 139-139 (2007)
Andrey Solovyev et al.
Journal of the American Chemical Society, 132(42), 15072-15080 (2010-10-05)
Boryl halide, carboxylate and sulfonate complexes of 1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene (dipp-Imd-BH(2)X, X = halide or sulfonate) have been prepared from the parent borane dipp-Imd-BH(3) by (1) substitution reactions with R-X (X = halide or sulfonate), (2) reactions with electrophiles (like I(2) or
Xaba, N.; Jaganyi, D.
Polyhedron, 28, 1145-1145 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service