Skip to Content
MilliporeSigma
All Photos(1)

Documents

261858

Sigma-Aldrich

2-Cyano-6-methoxybenzothiazole

99%

Synonym(s):

6-Methoxy-2-benzothiazolecarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6N2OS
CAS Number:
Molecular Weight:
190.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

129-131 °C (lit.)

solubility

chloroform: soluble 5%, clear, colorless to faintly yellow

SMILES string

COc1ccc2nc(sc2c1)C#N

InChI

1S/C9H6N2OS/c1-12-6-2-3-7-8(4-6)13-9(5-10)11-7/h2-4H,1H3

InChI key

DEWDWBYQOFXKIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Cyano-6-methoxybenzothiazole has been used in the synthesis of:
  • firefly luciferin via condensation with cysteine
  • luciferin β-glycosides, substrates for novel ultrasensitive enzyme assays
Luciferin intermediate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of luciferin glycosides as substrates for novel ultrasensitive enzyme assays.
Amess R, et al.
Carbohydrate Research, 205, 225-233 (1990)
Analogs of firefly luciferin.
White EH, et al.
The Journal of Organic Chemistry, 30(7), 2344-2348 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service