Skip to Content
MilliporeSigma
All Photos(1)

Documents

260118

Sigma-Aldrich

2-Amino-5-bromobenzoic acid

97%

Synonym(s):

5-Bromoanthranilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(Br)CO2H
CAS Number:
Molecular Weight:
216.03
Beilstein:
639028
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

213-215 °C (lit.)

application(s)

peptide synthesis

SMILES string

Nc1ccc(Br)cc1C(O)=O

InChI

1S/C7H6BrNO2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3H,9H2,(H,10,11)

InChI key

CUKXRHLWPSBCTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shuhua Cao et al.
Dalton transactions (Cambridge, England : 2003), 49(34), 11851-11858 (2020-07-24)
A bimetallic Cu(ii) complex as a novel antitumor chemodynamic therapy agent with glutathione (GSH) depletion properties is successfully synthesized and well characterized. In tumor cells, the Cu2+ ions of the complex are reduced to Cu+ ions by GSH and then
Rafael Gozalbes et al.
Bioorganic & medicinal chemistry, 18(19), 7078-7084 (2010-09-03)
Solubility plays a very important role in the selection of compounds for drug screening. In this context, a QSAR model was developed for predicting water solubility of drug-like compounds. First, a set of relevant parameters for establishing a drug-like chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service