Skip to Content
MilliporeSigma
All Photos(1)

Documents

252573

Sigma-Aldrich

Xanthydrol

98%

Synonym(s):

9-Hydroxyxanthene, 9-Xanthenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10O2
CAS Number:
Molecular Weight:
198.22
Beilstein:
10395
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

127-128 °C (lit.)

solubility

methanol: soluble 50 mg/mL, slightly hazy

storage temp.

2-8°C

SMILES string

OC1c2ccccc2Oc3ccccc13

InChI

1S/C13H10O2/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13/h1-8,13-14H

InChI key

JFRMYMMIJXLMBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xanthydrol is also known as 9H-xanthen-9-ol.

Application

Xanthydrol has been used as derivatization reagent in the determination of:
  • acrylamide in surface and drinking water by GC-MS method
  • urea by HPLC with fluorescence detection

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun-Hee Lim et al.
Journal of separation science, 36(18), 3059-3066 (2013-07-10)
A sensitive GC-MS method has been established for the determination of acrylamide in surface and drinking water based on derivatization with xanthydrol. Deuterated acrylamide (acrylamide-d3 ) was chosen as the internal standard for analyzing the water sample. The derivatization of
Kumiko Yamazaki et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 29(5), 705-715 (2012-01-20)
A novel GC-MS method was developed for the determination of acrylamide, which is applicable to a variety of processed foods, including potato snacks, corn snacks, biscuits, instant noodles, coffee, soy sauces and miso (fermented soy bean paste). The method involves
Shona Clark et al.
Journal of chromatography. A, 1161(1-2), 207-213 (2007-06-16)
A high-performance liquid chromatography (HPLC) method for the determination of urea that incorporates automated derivatisation with xanthydrol (9H-xanthen-9-ol) is described. Unlike the classic xanthydrol approach for the determination of urea, which involves the precipitation of dixanthylurea (N,N'-di-9H-xanthen-9-ylurea), the derivatisation procedure
P V Biles et al.
Journal of AOAC International, 81(6), 1155-1161 (1998-12-16)
Contamination of food and food packaging material by rodent urine is evidence of insanitary conditions. Urea from rodent urine is used as a chemical indicator of contamination. The limit of detection of the xanthydrol/urea AOAC Method 959.14 by formation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service