Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

251658

Sigma-Aldrich

3-Sulfopropyl methacrylate potassium salt

98%

Synonym(s):

Methacrylic Acid 3-Sulfopropyl Ester Potassium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2(CH2)3SO3K
CAS Number:
Molecular Weight:
246.32
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

mp

295 °C (dec.) (lit.)

SMILES string

[K+].CC(=C)C(=O)OCCCS([O-])(=O)=O

InChI

1S/C7H12O5S.K/c1-6(2)7(8)12-4-3-5-13(9,10)11;/h1,3-5H2,2H3,(H,9,10,11);/q;+1/p-1

InChI key

PNOXUQIZPBURMT-UHFFFAOYSA-M

Application

  • Antifouling and flux enhancement of reverse osmosis membrane by grafting poly (3-sulfopropyl methacrylate) brushes: This study explores the enhancement of antifouling properties in reverse osmosis membranes through grafting, addressing significant issues in membrane-based water treatment technologies (Mushtaq et al., 2021).
  • Effect of cation alkyl chain length on 3-sulfopropylmethacrylate-based draw solutes having lower critical solution temperature: This article explores the physicochemical properties of ionic liquids derived from 3-sulfopropyl methacrylate, important for developing new materials with specific thermal and solubility properties (Moon et al., 2023).
  • Transpicuous-cum-fouling resistant copolymers of 3-sulfopropyl methacrylate and methyl methacrylate for optronics applications in aquatic medium: Discusses the development of copolymers for optical applications, particularly in environments where fouling resistance is crucial, relevant for material science (Mushtaq et al., 2020).
  • A general approach for construction of asymmetric modification membranes for gated flow nanochannels: This research introduces a method for modifying membranes with nanochannels using 3-sulfopropyl methacrylate, important for creating selective transport pathways in sensor and separation technologies (Ma et al., 2014).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed Salama
International journal of biological macromolecules, 106, 940-946 (2017-08-24)
A novel superadsorbent anionic hydrogel was synthesized by grafting of poly (3-sulfopropyl methacrylate), P(SPMA), onto carboxymethyl cellulose (CMC). CMC-g-P(SPMA) superadsorbent hydrogel was applied as an efficient and sustainable adsorbent to remove methylene blue (MB) from waste water. Batch adsorption experiments
Yu Zhang et al.
Biomaterials science, 5(12), 2493-2500 (2017-11-09)
The current paper reports a novel model of a marine antibiofouling surface based on polymer brushes on a wrinkled silicone elastomer. Polymer brushes (POEGMA and PSPMA) were grafted via surface-initiated atom transfer radical polymerization (SI-ATRP). Successful grafting was verified with
Antoine Venault et al.
Colloids and surfaces. B, Biointerfaces, 151, 372-383 (2017-01-08)
This work discusses the impact of the charge bias and the hydrophilicity on the human blood compatibility of pseudozwitterionic biomaterial gels. Four series of hydrogels were prepared, all containing negatively-charged 3-sulfopropyl methacrylate (SA), and either acrylamide, N-isopropylacrylamide, 2-dimethylaminoethyl methacrylate (DMAEMA)
Armin Eraghi Kazzaz et al.
Journal of colloid and interface science, 561, 231-243 (2019-12-13)
Amphoteric polymers have tremendous applications in colloidal systems. Despite the enormous availability of lignin, the generation of amphoteric lignin polymer has yet to be discovered. In this work, amphoteric lignin was synthesized in an industrially attractive aqueous system via integrating
Yi-Li Lin et al.
Environmental science and pollution research international, 26(33), 34073-34084 (2018-09-29)
A commercially available nanofiltration membrane, NF90, was modified using an in situ concentration polarization-enhanced radical graft polymerization method to improve the organic fouling resistance as well as the removal of pharmaceutical and personal care products (PPCPs), including ibuprofen (IBU), carbamazepine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service