Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

250066

Sigma-Aldrich

Aluminum bromide solution

anhydrous, 1.0 M in dibromomethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AlBr3
CAS Number:
Molecular Weight:
266.69
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
liquid

grade

anhydrous

vapor pressure

1 mmHg ( 81.3 °C)

form

liquid

reaction suitability

reagent type: catalyst
core: aluminum

concentration

1.0 M in dibromomethane

density

2.522 g/mL at 25 °C

SMILES string

Br[Al](Br)Br

InChI

1S/Al.3BrH/h;3*1H/q+3;;;/p-3

InChI key

PQLAYKMGZDUDLQ-UHFFFAOYSA-K

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service