Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

249939

Sigma-Aldrich

Bicyclo[2.2.1]hepta-2,5-diene-rhodium(I) chloride dimer

96%

Synonym(s):

[Rh(nbd)Cl]2, dichlorobis(norbornadiene)dirhodium, 2,5-Norbornadiene-rhodium(I) chloride dimer, Chloro(2,5-norbornadiene)rhodium(I) dimer, Rhodium(I) chloride 2,5-Norbornadiene complex dimer, [Rh(nbd)Cl]2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16Cl2Rh2
CAS Number:
Molecular Weight:
460.99
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

powder

reaction suitability

core: rhodium
reagent type: catalyst

SMILES string

Cl[Rh].Cl[Rh].[CH]1[CH]C2[CH][CH]C1C2.[CH]3[CH]C4[CH][CH]C3C4

InChI

1S/2C7H8.2ClH.2Rh/c2*1-2-7-4-3-6(1)5-7;;;;/h2*1-4,6-7H,5H2;2*1H;;/q;;;;2*+1/p-2/t2*6-,7+;;;;

InChI key

RXDWVIOULPVOEO-SEGRDSFWSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.

Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.

Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.

Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service