Skip to Content
MilliporeSigma
All Photos(1)

Documents

247189

Sigma-Aldrich

4-Methoxyphenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4NCS
CAS Number:
Molecular Weight:
165.21
Beilstein:
606967
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.653 (lit.)

bp

280-281 °C (lit.)

mp

18 °C (lit.)

density

1.196 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(cc1)N=C=S

InChI

1S/C8H7NOS/c1-10-8-4-2-7(3-5-8)9-6-11/h2-5H,1H3

InChI key

VRPQCVLBOZOYCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

4-Methoxyphenyl isothiocyanate has been used in the synthesis of:
  • 3-benzyl-2-(4-methoxyphenyl)-3H-[1,2,4]triazolo-[5,1-b]quinazolin-9-one
  • 2-{[4-amino-3-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetyl}-N-arylhydrazinecarbothioamides

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veerachamy Alagarsamy et al.
Bioorganic & medicinal chemistry, 15(10), 3457-3462 (2007-03-30)
A series of 3-benzyl-2-substituted-3H-[1,2,4]triazolo[5,1-b]quinazolin-9-ones have been synthesized by the cyclocondensation of 3-amino-2-benzylamino-3H-quinazolin-4-one with a variety of one-carbon donors. The starting material 3-amino-2-benzylamino-3H-quinazolin-4-one was synthesized from methyl anthranilate by a novel innovative route. The title compounds were evaluated for their in
M K Dewanjee et al.
Bioconjugate chemistry, 2(4), 195-200 (1991-07-01)
Radioactive and nonradioactive oligonucleotide (ON) probes have been used in a variety of studies of in vitro hybridization for locations of specific genes and determination of the level of mRNA transcription in activated and proliferative cells. Nuclease-resistant phosphorothioate derivatives of
María C Soraires Santacruz et al.
Bioorganic & medicinal chemistry, 25(15), 4055-4063 (2017-06-11)
A series of N
Linhua Huang et al.
Molecules (Basel, Switzerland), 25(11) (2020-06-03)
Due to the side effects of obesity medications, many studies have focused on the natural products used in the daily diet to control weight. Moringa seed pods and leaves are widely used as vegetables or diet supplements due to the
Ahmet Demirbas et al.
European journal of medicinal chemistry, 44(7), 2896-2903 (2009-01-27)
4-Amino-2-[(5-arylamino-4,5-dihydro-1,3,4-thiadiazol-2-yl)methyl]-5-(4-methylphenyl)-2,4-dihydro-3H-1,2,4-triazol-3-ones (3a-c) were obtained in acidic media via the formation of 2-[(4-amino-3-aryl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl)acetyl]-N-arylhydrazinecarbothioamides (2a-c), and then, compound 3b was converted to methylated derivative, 4. The basic treatment of carbothioamide derivatives, 2a-c, afforded 4-amino-2-[(4-aryl-5-sulphanyl-4H-1,2,4-triazol-3-yl)methyl]-5-(4-methylphenyl)-2,4-dihydro-3H-1,2,4-triazol-3-ones (5a-c). The alkylation reactions of compounds 4H-1,2,4-triazol-3-ylmethyl-5-(4-methylphenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service