Skip to Content
MilliporeSigma
All Photos(3)

Documents

247014

Sigma-Aldrich

Boron trichloride methyl sulfide complex

99%

Synonym(s):

Dimethyl sulfide-trichloroborane, Trichloroborane-methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S · BCl3
CAS Number:
Molecular Weight:
179.30
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

mp

88-90 °C (lit.)

SMILES string

CSC.ClB(Cl)Cl

InChI

1S/C2H6BCl3S/c1-7(2)3(4,5)6/h1-2H3

InChI key

YNDYUDLLOGOQNW-UHFFFAOYSA-N

General description

Boron trichloride in convenient solid form.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service