Skip to Content
MilliporeSigma

242551

1,5-Pentanedithiol

96%

Synonym(s):

Pentamethylene dimercaptan

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HS(CH2)5SH
CAS Number:
Molecular Weight:
136.28
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
213-194-9
Beilstein/REAXYS Number:
1732335
MDL number:
Assay:
96%
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

96%

form

liquid

refractive index

n20/D 1.519 (lit.)

bp

107-108 °C/15 mmHg (lit.)

mp

−72 °C (lit.)

density

1.016 g/mL at 25 °C (lit.)

functional group

thiol

SMILES string

SCCCCCS

InChI

1S/C5H12S2/c6-4-2-1-3-5-7/h6-7H,1-5H2

InChI key

KMTUBAIXCBHPIZ-UHFFFAOYSA-N

Application

1,5-Pentanedithiol was used as a cross-linker in the preparation of colloidal Au/Ag multilayer films. It was also used as derivatization reagent in determination of organoarsenic compounds in fish by GC-MS analysis.


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

203.0 °F - closed cup

Flash Point (°C)

95 °C - closed cup

PPE (personal protective equipment)

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library