Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

235954

Sigma-Aldrich

Tetrapropylammonium iodide

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)4N(I)
CAS Number:
Molecular Weight:
313.26
Beilstein:
3567386
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

solid

mp

280-285 °C (dec.) (lit.)

SMILES string

[I-].CCC[N+](CCC)(CCC)CCC

InChI

1S/C12H28N.HI/c1-5-9-13(10-6-2,11-7-3)12-8-4;/h5-12H2,1-4H3;1H/q+1;/p-1

InChI key

GKXDJYKZFZVASJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vidhya Selvanathan et al.
Polymers, 12(3) (2020-03-04)
This work is a pioneer attempt to fabricate quasi-solid dye-sensitized solar cell (QSDDSC) based on organosoluble starch derivative. Rheological characterizations of the PhSt-HEC blend based gels exhibited viscoelastic properties favorable for electrolyte fabrication. From amplitude sweep and tack test analyses
Alexey A Sysoev et al.
European journal of mass spectrometry (Chichester, England), 22(6), 289-296 (2016-12-03)
An ion mobility time-of-flight mass spectrometry (IM-TOFMS)-based method has been preliminarily investigated for the identification of triphenylmethane ballpoint pen dyes on paper. The dyes were sampled from one-year-old ballpoint pen ink entries. The entries were written on paper documents stored
Hui Wang et al.
Physical chemistry chemical physics : PCCP, 17(32), 20636-20646 (2015-07-24)
The (13)C NMR chemical shift moving upfield indicates the main model of π-holeX(-) bond between cyanuric chloride/1,3,5-triazine (3ClN/3N), which possess both the π-hole and σ-hole, and X(-). (13)C NMR and UV absorption titration in acetonitrile confirmed that the bonding abilities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service