Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

233323

Sigma-Aldrich

Acetonitrile-d3

"100%", 99.96 atom % D

Synonym(s):

Methyl-d3 cyanide, Trideuteroacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CN
CAS Number:
Molecular Weight:
44.07
Beilstein:
1740230
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

isotopic purity

99.96 atom % D

Assay

99% (CP)

form

liquid

expl. lim.

3.0-16 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.0200% water
water

refractive index

n20/D 1.341 (lit.)

bp

80.7 °C (lit.)

mp

-46 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])C#N

InChI

1S/C2H3N/c1-2-3/h1H3/i1D3

InChI key

WEVYAHXRMPXWCK-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetonitrile-d3 is (Trideuteroacetonitrile, CD3CN) is a deuterated form of acetonitrile. It is 100% isotopically enriched NMR (nuclear magnetic resonance) solvent. It is widely employed in high resolution NMR studies due to its high chemical and isotopic purity.

Application

Acetonitrile-d3 used as a solvent:
  • During the synthesis of 2- phenylindole to determined yield of the by NMR spectroscopy.
  • For 1H NMR (proton nuclear magnetic resonance) spectral studies.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bryan J Leong et al.
Plant physiology, 183(3), 915-924 (2020-05-02)
Plants make many biologically active, specialized metabolites, which vary in structure, biosynthesis, and the processes they influence. An increasing number of these compounds are documented to protect plants from insects, pathogens, or herbivores or to mediate interactions with beneficial organisms
Márk Szabó et al.
Chirality, 32(4), 446-456 (2020-02-08)
Dirhodium complexes bearing N-substituted chiral amino acid ligands are investigated. These complexes have an unusual twisted paddlewheel structure, showing inherent chirality. We would like to demonstrate that parallel application of chiroptical spectroscopic methods (ECD and VCD) and NMR spectroscopy combined
Magnetic Resonance Studies of Anisotropic Molecular Rotation in Liquid Acetonitrile-d3.
Bopp TT.
J. Chem. Phys. , 47(9), 3621-3626 (1967)
[Cp*IrCl2]2-catalysed cyclization of 2-alkynylanilines into indoles.
Kumaran E and Leong WK.
Tetrahedron Letters, 55(40), 5495-5498 (2014)
Pressure dependencies of rotational, translational, and viscous friction coefficients in water-d2, acetonitrile-d3, acetonitrile, chloroform, and benzene.
Wakai C and Nakahara M.
J. Chem. Phys. , 100(11), 8347-8358 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service