Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

229644

Sigma-Aldrich

Gallium(III) nitrate hydrate

crystals and lumps, 99.999% trace metals basis

Synonym(s):

Gallium trinitrate hydrate, Hydrated gallium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ga(NO3)3 · xH2O
CAS Number:
Molecular Weight:
255.74 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
crystals and lumps

Quality Level

Assay

99.999% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: gallium

impurities

≤20.0 ppm Trace Rare Earth Analysis

mp

110 °C (dec.) (lit.)

SMILES string

O.[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ga.3NO3.H2O/c;3*2-1(3)4;/h;;;;1H2/q+3;3*-1;

InChI key

YVFORYDECCQDAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Gallium (III) nitrate hydrate was used as one of the precursors for the preparation of nanoporous TiO2–Ga2O3 binary metal oxide films and powders by sol gel method. These nanostructures can find applications in optical electronics, gas sensors etc.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterisation of nanostructural TiO2?Ga2O3 binary oxides with high surface area derived form particulate sol?gel route.
Mohammadi MR, et al.
J. Mater. Sci., 42(13), 4976-4986 (2007)
Comparison of single and binary oxide sol?gel gas sensors based on titania
Mohammadi MR, et al.
Solid State Sciences, 10(7), 884-893 (2008)
Melnikov P, et al.
Journal of Thermal Analysis and Calorimetry, 107(3), 1117-1121 (2012)
Matthias Metzger et al.
Analytical and bioanalytical chemistry, 411(19), 4647-4660 (2019-03-09)
The introduction of fluorine into organic molecules leads to new chemical/physical properties. Especially in the field of pharmaceutical as well as technical applications, fluorinated organic substances gain in importance. The OECD identified and categorized 4730 per- and polyfluoroalkyl substances-related CAS
Charlotte Mellier et al.
Acta biomaterialia, 24, 322-332 (2015-06-16)
Different possible options were investigated to combine an apatitic calcium phosphate cement with gallium ions, known as bone resorption inhibitors. Gallium can be either chemisorbed onto calcium-deficient apatite or inserted in the structure of β-tricalcium phosphate, and addition of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service