Skip to Content
MilliporeSigma
All Photos(1)

Documents

229156

Sigma-Aldrich

6-Methoxyquinoline N-oxide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

102-104 °C (lit.)

SMILES string

COc1ccc2[n+]([O-])cccc2c1

InChI

1S/C10H9NO2/c1-13-9-4-5-10-8(7-9)3-2-6-11(10)12/h2-7H,1H3

InChI key

BWEGRKPOJXNZSK-UHFFFAOYSA-N

General description

The standard molar enthalpy of formation of 6-methoxyquinoline N-oxide was studied using static-bomb calorimetry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Energetics of 6-methoxyquinoline and 6-methoxyquinoline< i> N</i>-oxide: the dissociation enthalpy of the (N?O) bond.
Ribeiro da Silva MD, et al.
The Journal of Chemical Thermodynamics, 35(7), 1093-1100 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service