Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

227277

Sigma-Aldrich

Silver benzoate

99%

Synonym(s):

Silver benzoate (AgOBz), Silver(1+) benzoate, Silver(I) benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2Ag
CAS Number:
Molecular Weight:
228.98
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder or crystals

solubility

cold water: 385 part(lit.)
alcohol: slightly soluble(lit.)
hot water: soluble (more soluble)(lit.)

functional group

phenyl

SMILES string

[Ag]OC(=O)c1ccccc1

InChI

1S/C7H6O2.Ag/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1

InChI key

CLDWGXZGFUNWKB-UHFFFAOYSA-M

General description

Silver benzoate along with 7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene (MTBD) is an efficient catalytic system for the reaction of carbon dioxide with various ketones.

Application

Silver benzoate was used in the synthesis of triphenyltinbenzoate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M K Choudhury et al.
Indian journal of pharmaceutical sciences, 72(4), 531-533 (2011-01-11)
Triphenyltinbenzoate was synthesized using triphenyltinchloride and silver benzoate prepared from sodium benzoate. The structure of the synthetic compound was elucidated by spectral and C, H analysis. The antibacterial activities of the organotin compound were determined against four bacteria namely Escherichia
Satoshi Kikuchi et al.
Angewandte Chemie (International ed. in English), 51(28), 6989-6992 (2012-06-08)
A catalytic amount of silver benzoate with 7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene (MTBD) was an effective catalytic system for the reaction of carbon dioxide with various ketones containing an alkyne group at an appropriate position. These reactions afforded the corresponding γ-lactone derivatives in good

Articles

Copper metal deposition processes are an essential tool for depositing interconnects used in microelectronic applications, giving group 11 (coinage metals: Copper, Silver, and Gold) an important place in atomic layer deposition (ALD) process development.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service