Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

226181

Sigma-Aldrich

Chloro(chloromethyl)dimethylsilane

98%

Synonym(s):

(Chloromethyl)dimethylchlorosilane, CMDMCS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2Si(CH3)2Cl
CAS Number:
Molecular Weight:
143.09
Beilstein:
1731933
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.437 (lit.)

bp

114 °C/752 mmHg (lit.)

density

1.086 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

C[Si](C)(Cl)CCl

InChI

1S/C3H8Cl2Si/c1-6(2,5)3-4/h3H2,1-2H3

InChI key

ITKVLPYNJQOCPW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aldrichimica Acta, 14, 40-40 (1981)
The Journal of Organic Chemistry, 57, 6552-6552 (1992)
Tetrahedron Letters, 34, 1449-1449 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service