Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

225924

Sigma-Aldrich

LS-Selectride® solution

1.0 M in THF

Synonym(s):

Lithium trisiamylborohydride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiB[CH(CH3)CH(CH3)2]3H
CAS Number:
Molecular Weight:
232.18
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

1.0 M in THF

density

0.903 g/mL at 25 °C

SMILES string

[Li+].CC(C)C(C)[BH-](C(C)C(C)C)C(C)C(C)C

InChI

1S/C15H34B.Li/c1-10(2)13(7)16(14(8)11(3)4)15(9)12(5)6;/h10-16H,1-9H3;/q-1;+1

InChI key

DGIADQRDWIBVOW-UHFFFAOYSA-N

Application

Reactant for:
  • Nucleophilic addition reactions
  • Conjugate reduction-initiated cyclization reactions

Legal Information

Selectride is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service