Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

225908

Sigma-Aldrich

Allylmagnesium chloride solution

2.0 M in THF

Synonym(s):

Allyl magnesium chloride, Allylchloromagnesium, Allylmagnesium chloride, Chloro-2-propen-1-ylmagnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2MgCl
CAS Number:
Molecular Weight:
100.83
Beilstein:
3587316
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in THF

density

0.995 g/mL at 25 °C

SMILES string

Cl[Mg]CC=C

InChI

1S/C3H5.ClH.Mg/c1-3-2;;/h3H,1-2H2;1H;/q;;+1/p-1

InChI key

PLYLAFITHJUEGX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Allylmagnesiumchloride solution is an organic compound commonly used as a Grignard reagent inchemical reactions. It is used for the allylation of carbonyl compounds, such as aldehydes and ketones.

Application

Allylmagnesium chloride solution is used as a Grignard reagent in the preparation of:
  • Antipsychotic clopixol.
  • Polysilane–crown ether polymer (PSEC) using hexachloroplatinic acid as a catalyst.
  • 1,7-disubstituted-1,2,3,4-tetrahydroisoquinolinesfrom 3,4-dihydroisoquinoline.

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup - Solvent

Flash Point(C)

-17 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, In Vitro and In Silico Analysis of New Oleanolic Acid and Lupeol Derivatives against Leukemia Cell Lines: Involvement of the NF-?B Pathway
Fontana G, et al.
International Journal of Molecular Sciences, 6594-6594 (2022)
Tetrahedron Letters, 47, 9207-9207 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service