Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

225371

Sigma-Aldrich

1,4-Dichloro-2-nitrobenzene

99%

Synonym(s):

2,5-Dichloronitrobenzene, Nitro-p-dichlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NO2
CAS Number:
Molecular Weight:
192.00
Beilstein:
778109
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.6 (vs air)

vapor pressure

<0.1 mmHg ( 25 °C)

Assay

99%

form

solid

expl. lim.

9.2 %

bp

266-269 °C (lit.)

mp

52-54 °C (lit.)

SMILES string

[O-][N+](=O)c1cc(Cl)ccc1Cl

InChI

1S/C6H3Cl2NO2/c7-4-1-2-5(8)6(3-4)9(10)11/h1-3H

InChI key

RZKKOBGFCAHLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

275.0 °F

Flash Point(C)

135 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z C Jin et al.
Zhonghua yu fang yi xue za zhi [Chinese journal of preventive medicine], 25(5), 292-295 (1991-09-01)
The plasmid pSK1002 carrying the fused gene umuC'-'lacZ could increase the number of revertants induced by methyl methanesulfonate (MMS) and 4-nitroquinoline-1-oxide (4NQO) in S. typhimurium LT2 TA 1535(his-). Maximum revertants were 4.35 and 3.96 times that of the controls without
Z C Jin et al.
Mutation research, 264(1), 1-5 (1991-09-01)
The plasmid pSK1002 (umuC'-'lacZ) could increase the number of revertants induced by methyl methanesulfonate (MMS) and 4-nitroquinoline-N-oxide (4NQO) in Salmonella typhimurium TA 1535 (his-). The values induced by N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) were not different irrespective of the presence or absence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service